Assay Details
| Gene Symbol: KPNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002268.4 | NP_002259.1 | 17 | 8279 | 77 | |
| GenBank mRNA | AF038199.1 | - | 1 | 420 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KPNA4| Gene Symbol: KPNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002268.4 | NP_002259.1 | 17 | 8279 | 77 | |
| GenBank mRNA | AF038199.1 | - | 1 | 420 | 77 | |
Set Membership: |
|