Assay Details
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034845.2 | NP_001030017.2 | 5 - 6 | 1285 | 85 | |
XM_011531993.2 | XP_011530295.1 | 4 - 5 | 623 | 85 | ||
XM_017008243.1 | XP_016863732.1 | 5 - 6 | 2019 | 85 | ||
XM_017008244.1 | XP_016863733.1 | 4 - 5 | 1391 | 85 | ||
GenBank mRNA | AJ626725.1 | - | 4 - 5 | 498 | 85 | |
AK295464.1 | - | 4 - 5 | 702 | 85 | ||
BC047551.1 | - | 5 - 6 | 1206 | 5301536 | 85 | |
GU220060.1 | - | 4 - 5 | 549 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000457021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000504379.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000506823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000508122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000511251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000513061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | ENST00000616459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009318.hg.1 | NM_001034845.2 | Refseq |
