Assay Details
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011531702.2 | XP_011530004.1 | 2 - 3 | 1263 | 96 | |
XM_017007839.1 | XP_016863328.1 | 2 - 3 | 454 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011531702.2, XM_017007839.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000359079.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394684.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000394686.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000503862.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000506993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | ENST00000515332.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136257.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_001136258.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008345.hg.1 | NM_152621.5 | Refseq |
