Assay Details
Gene Symbol: CCDC110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011531827.2 | XP_011530129.1 | 6 - 7 | 2527 | 65 | |
XM_011531828.2 | XP_011530130.1 | 6 - 7 | 2604 | 65 | ||
XM_011531829.2 | XP_011530131.1 | 5 - 6 | 2471 | 65 | ||
XM_017007987.1 | XP_016863476.1 | 5 - 6 | 2512 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000307588.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000393540.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000504020.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000506962.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000507501.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000508538.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | ENST00000510617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_001145411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012640.hg.1 | NM_152775.3 | Refseq |
