Assay Details
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006835.2 | NP_006826.1 | 5 - 6 | 1007 | 67 | |
GenBank mRNA | AB209766.1 | - | 5 - 6 | 1009 | 67 | |
AF135162.1 | - | 5 - 6 | 662 | 67 | ||
AK222931.1 | - | 4 - 5 | 459 | 67 | ||
AK301111.1 | - | 4 - 5 | 868 | 67 | ||
AK312632.1 | - | 7 - 8 | 877 | 67 | ||
BC000420.2 | - | 5 - 6 | 1006 | 2964432 | 67 | |
BC004975.1 | - | 5 - 6 | 1006 | 2957878 | 67 | |
BT020019.1 | - | 4 - 5 | 463 | 67 | ||
CR541783.1 | - | 4 - 5 | 463 | 67 | ||
D50310.1 | - | 4 - 5 | 463 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
