| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000513733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
ENST00000513733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000286794.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000511542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000513733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
ENST00000513733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011134.hg.1 |
NM_032693.2 |
Refseq |