Assay Details
| Gene Symbol: PPM1K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006714111.3 | XP_006714174.1 | 1 - 2 | 485 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006714111.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |

PPM1K
Genomic Map