Assay Details
| Gene Symbol: EGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178130.2 | NP_001171601.1 | 3 - 4 | 961 | 136 | |
| NM_001178131.2 | NP_001171602.1 | 3 - 4 | 961 | 136 | ||
| NM_001963.5 | NP_001954.2 | 3 - 4 | 961 | 136 | ||
| XM_005262796.2 | XP_005262853.1 | 3 - 4 | 962 | 136 | ||
| XM_011531707.1 | XP_011530009.1 | 4 - 5 | 611 | 136 | ||
| XM_017007845.1 | XP_016863334.1 | 3 - 4 | 986 | 136 | ||
| XM_017007846.1 | XP_016863335.1 | 3 - 4 | 986 | 136 | ||
| XM_017007847.1 | XP_016863336.1 | 3 - 4 | 986 | 136 | ||
| XM_017007848.1 | XP_016863337.1 | 3 - 4 | 986 | 136 | ||
| XM_017007849.1 | XP_016863338.1 | 3 - 4 | 986 | 136 | ||
| XM_017007850.1 | XP_016863339.1 | 3 - 4 | 986 | 136 | ||
| XM_017007851.1 | XP_016863340.1 | 3 - 4 | 986 | 136 | ||
| XM_017007852.1 | XP_016863341.1 | 3 - 4 | 962 | 136 | ||
| XM_017007853.1 | XP_016863342.1 | 3 - 4 | 986 | 136 | ||
| XM_017007854.1 | XP_016863343.1 | 3 - 4 | 986 | 136 | ||
| XM_017007855.1 | XP_016863344.1 | 3 - 4 | 986 | 136 | ||
| GenBank mRNA | AK299306.1 | - | 3 - 4 | 963 | 136 | |
| BC093731.1 | - | 3 - 4 | 604 | 7939576 | 136 | |
| BC113461.1 | - | 3 - 4 | 604 | 8322513 | 136 | |
| BC143357.1 | - | 3 - 4 | 604 | 136 | ||
| X04571.1 | - | 3 - 4 | 945 | 136 | ||
More Information
Set Membership: |
|

EGF
Genomic Map