Assay Details
| Gene Symbol: PPM1K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152542.4 | NP_689755.3 | 7 | 5954 | 131 | |
| XM_006714111.3 | XP_006714174.1 | 7 | 6015 | 131 | ||
| XM_017007803.1 | XP_016863292.1 | 7 | 5602 | 131 | ||
| XM_017007804.1 | XP_016863293.1 | 8 | 6142 | 131 | ||
| XM_017007805.1 | XP_016863294.1 | 9 | 6142 | 131 | ||
| XM_017007806.1 | XP_016863295.1 | 7 | 5475 | 131 | ||
| XM_017007807.1 | XP_016863296.1 | 6 | 5193 | 131 | ||
| GenBank mRNA | AK092450.1 | - | 1 | 1109 | 131 | |
More Information
Set Membership: |
|

PPM1K
Genomic Map