Assay Details
| Gene Symbol: ARAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007698.1 | XP_016863187.1 | 35 - 36 | 5915 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARAP2| Gene Symbol: ARAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007698.1 | XP_016863187.1 | 35 - 36 | 5915 | 93 | |
Set Membership: |
|