Assay Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020846.1 | NP_065897.1 | 1 | 297 | 112 | |
| XM_005248121.2 | XP_005248178.1 | 1 | 413 | 112 | ||
| GenBank mRNA | AB040891.1 | - | 1 | 267 | 112 | |
| AK291006.1 | - | 1 | 427 | 112 | ||
| BC006139.1 | - | 1 | 322 | 112 | ||
| BC012067.1 | - | 1 | 322 | 112 | ||
| BC040993.1 | - | 1 | 576 | 5272534 | 112 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |

SLAIN2
Genomic Map