Assay Details
| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY250185.1 | - | 1 | 218 | 57 | |
| BC101193.2 | - | 1 | 218 | 57 | ||
| BC101194.2 | - | 1 | 218 | 57 | ||
| BC101195.1 | - | 1 | 218 | 57 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000318186.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000451762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | ENST00000624443.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008025.hg.1 | NR_015359.1 | Refseq |

WDFY3-AS2
Genomic Map