Assay Details
Gene Symbol: LOC105369192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134236.1 | 3 - 4 | 329 | 96 | ||
NR_134237.1 | 2 - 3 | 253 | 96 | |||
GenBank mRNA | BC027846.1 | - | 3 - 4 | 317 | 96 | |
DA132869.1 | - | 2 - 3 | 253 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
