Assay Details
Gene Symbol: BEND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159547.1 | NP_001153019.1 | 6 | 8232 | 104 | |
NM_207406.3 | NP_997289.2 | 6 | 8303 | 104 | ||
XM_017008185.1 | XP_016863674.1 | 5 | 7960 | 104 | ||
GenBank mRNA | AB126828.1 | - | 6 | 8303 | 104 | |
AB126829.1 | - | 6 | 8232 | 104 | ||
AK092951.1 | - | 1 | 1920 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
