Assay Details
| Gene Symbol: PLA2G12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030821.4 | NP_110448.2 | 4 | 3496 | 71 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
biological_process
lipid catabolic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipase A2 activity
phosphatidic acid biosynthetic process
biological_process
lipid catabolic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipase A2 activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |

PLA2G12A
Genomic Map