Assay Details
| Gene Symbol: LOC728095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098705.1 | - | 1 - 2 | 122 | 111 | |
| DB036245.1 | - | 1 - 2 | 138 | 111 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |

LOC728095
Genomic Map