Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317981.1 | NP_001304910.1 | 1 - 2 | 73 | 70 | |
| GenBank mRNA | BI838595.1 | - | 73 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001317981.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
structural constituent of ribosome
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
structural constituent of ribosome
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000265100.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000519974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | ENST00000521476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009478.hg.1 | NM_016093.3 | Refseq |

RPL26L1
Genomic Map