Assay Details
Gene Symbol: ERGIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR749461.1 | - | 1 - 2 | 123 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523366.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523366.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000326654.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000393784.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000518247.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519796.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000519860.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520399.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000520642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000521392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523366.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523366.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | ENST00000523650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009470.hg.1 | NM_001031711.2 | Refseq |
