Assay Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098272.2 | NP_001091742.1 | 2 - 3 | 216 | 64 | |
| NM_001324220.1 | NP_001311149.1 | 2 - 3 | 236 | 64 | ||
| NM_001324222.1 | NP_001311151.1 | 2 - 3 | 216 | 64 | ||
| NM_001324224.1 | NP_001311153.1 | 2 - 3 | 236 | 64 | ||
| XM_017009412.1 | XP_016864901.1 | 2 - 3 | 296 | 64 | ||
| GenBank mRNA | AK315593.1 | - | 2 - 3 | 144 | 64 | |
| BC000297.2 | - | 2 - 3 | 95 | 64 | ||
| BJ995153.1 | - | 2 - 3 | 200 | 64 | ||
| M15802.1 | - | 2 - 3 | 112 | 64 | ||
| X66435.1 | - | 2 - 3 | 112 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK315593.1, NM_001098272.2, NM_001324220.1, NM_001324222.1, NM_001324224.1, X66435.1, XM_017009412.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
liver development
lipid metabolic process
cholesterol biosynthetic process
brain development
isoprenoid biosynthetic process
male gonad development
response to low light intensity stimulus
response to purine-containing compound
response to vitamin E
response to drug
response to tellurium ion
response to lipoprotein particle
cellular response to follicle-stimulating hormone stimulus
cellular response to cholesterol
hydroxymethylglutaryl-CoA synthase activity
drug binding
isomerase activity
protein homodimerization activity
organic acid binding
liver development
lipid metabolic process
cholesterol biosynthetic process
brain development
isoprenoid biosynthetic process
male gonad development
response to low light intensity stimulus
response to purine-containing compound
response to vitamin E
response to drug
response to tellurium ion
response to lipoprotein particle
cellular response to follicle-stimulating hormone stimulus
cellular response to cholesterol
hydroxymethylglutaryl-CoA synthase activity
drug binding
isomerase activity
protein homodimerization activity
organic acid binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000511774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000511774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012146.hg.1 | ENST00000400943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012146.hg.1 | ENST00000400943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000325110.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000433297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000507293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000508319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000511774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000511774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | ENST00000514610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_001098272.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010635.hg.1 | NM_002130.7 | Refseq |

HMGCS1
Genomic Map