Assay Details
| Gene Symbol: TMEM161B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DA782199.1 | - | 2 - 3 | 271 | 84 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000496733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000501869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000504922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000506584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000507736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000507736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000507736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000507736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000507736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000508015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000508015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000508015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000508015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000508015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000510087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000512724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513011.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513011.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513011.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513011.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513011.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | ENST00000513694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039995.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039995.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039995.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039995.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_039995.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008026.hg.1 | NR_105020.1 | Refseq |

TMEM161B-AS1
Genomic Map