Assay Details
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011543100.2 | XP_011541402.1 | 1 - 2 | 393 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011543100.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
cell proliferation
positive regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA-directed RNA polymerase activity
regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
cell proliferation
positive regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA-directed RNA polymerase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000399107.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503373.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000503973.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000504930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000505345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000512239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000514483.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | ENST00000622335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008054.hg.1 | NM_006467.2 | Refseq |
