| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000625713.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000629389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
ENST00000630823.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008065.hg.1 |
NR_027435.1 |
Refseq |