Assay Details
Gene Symbol: LOC100506639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BE545768.1 | - | 2 - 3 | 310 | 68 | |
CN309143.1 | - | 2 - 3 | 328 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000499046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000503599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000508795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | ENST00000514169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013166.hg.1 | NR_102752.1 | Refseq |
