| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000356555.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
ENST00000520773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012492.hg.1 |
NR_026867.1 |
Refseq |