Assay Details
| Gene Symbol: LINC00900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK309234.1 | - | 1 - 2 | 82 | 66 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000602803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | ENST00000602803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000499809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000501294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000514294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000535683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000537070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000602803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | ENST00000602803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012401.hg.1 | NR_034148.1 | Refseq |

LINC00900
Genomic Map