Assay Details
| Gene Symbol: C11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011542643.2 | XP_011540945.1 | 7 - 8 | 812 | 123 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |

C11orf65
Genomic Map