Assay Details
| Gene Symbol: TOLLIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029409.1 | 1 | 380 | 111 | ||
| GenBank mRNA | BC042808.1 | - | 1 | 261 | 111 | |
| BC066355.1 | - | 1 | 185 | 111 | ||
| DA359936.1 | - | 1 | 380 | 111 | ||
More Information
Set Membership: |
|

TOLLIP-AS1
Genomic Map