| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000227667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000375345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000433777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000470144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
ENST00000630701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009159.hg.1 |
NM_000040.2 |
Refseq |