Assay Details
Gene Symbol: LOC107984282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017017028.1 | XP_016872517.1 | 2 - 3 | 6707 | 114 | |
GenBank mRNA | AF086154.1 | - | 1 - 2 | 100 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
