Assay Details
Gene Symbol: GFRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005269696.1 | XP_005269753.1 | 1 - 2 | 94 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005269696.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
cell surface receptor signaling pathway
nervous system development
glial cell-derived neurotrophic factor receptor signaling pathway
positive regulation of GTPase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
glial cell-derived neurotrophic factor receptor activity
MAPK cascade
cell surface receptor signaling pathway
nervous system development
glial cell-derived neurotrophic factor receptor signaling pathway
positive regulation of GTPase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
glial cell-derived neurotrophic factor receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000355422.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000369236.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000439649.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | ENST00000490345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_001145453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_005264.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011923.hg.1 | NM_145793.4 | Refseq |
