Assay Details
Gene Symbol: CPXM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011539285.2 | XP_011537587.1 | 1 - 2 | 261 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011539285.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000241305.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000368854.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000481775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | ENST00000615851.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012108.hg.1 | NM_198148.2 | Refseq |
