Assay Details
Gene Symbol: LOC105376360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131187.1 | 1 - 2 | 160 | 116 | ||
GenBank mRNA | BC037918.1 | - | 1 - 2 | 162 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
