Assay Details
| Gene Symbol: ZNF503-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038223.1 | 3 | 1624 | 104 | ||
| NR_038224.1 | 3 | 1282 | 104 | |||
| NR_038225.1 | 4 | 1393 | 104 | |||
| NR_038226.1 | 4 | 1405 | 104 | |||
| GenBank mRNA | AK023445.1 | - | 3 | 1617 | 104 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000416398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000524517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000526759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000528121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000530606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000531808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | ENST00000533822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038223.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038224.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038225.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008094.hg.1 | NR_038226.1 | Refseq |

ZNF503-AS1
Genomic Map