| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000566977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000566977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568402.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
ENST00000581803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006469.hg.1 |
NR_049852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000219611.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000562370.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000565010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000566977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000566977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000567216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568402.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000568988.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
ENST00000581803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NM_005632.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006469.hg.1 |
NR_049852.1 |
Refseq |