Assay Details
| Gene Symbol: C5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271889.3 | XP_005271946.1 | 1 - 2 | 893 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005271889.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |

C5orf24
Genomic Map