| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000563101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000564167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010167.hg.1 |
ENST00000567048.1 |
Ensembl |