Assay Details
| Gene Symbol: HIST1H3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003529.2 | NP_003520.1 | 1 | 419 | 64 | |
| GenBank mRNA | BC066245.1 | - | 1 | 419 | 7002068 | 64 |
| BC066246.1 | - | 1 | 419 | 7002069 | 64 | |
| BC066247.1 | - | 1 | 419 | 7002070 | 64 | |
| BC067490.1 | - | 1 | 419 | 7002071 | 64 | |
| BC067491.1 | - | 1 | 419 | 7002072 | 64 | |
| BC069303.1 | - | 1 | 419 | 7262697 | 64 | |
More Information
Set Membership: |
|

HIST1H3A
Genomic Map