Assay Details
| Gene Symbol: HIST1H4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003540.3 | NP_003531.1 | 1 | 224 | 83 | |
| GenBank mRNA | BC069288.1 | - | 1 | 224 | 7262669 | 83 |
| BC093763.1 | - | 1 | 317 | 7939608 | 83 | |
| BC093765.1 | - | 1 | 317 | 7939610 | 83 | |
More Information
Set Membership: |
|

HIST1H4F
Genomic Map