| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000473369.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000473369.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000484554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007463.hg.1 |
ENST00000484554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377132.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000377136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000396792.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000473369.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000473369.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000484371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000484554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007463.hg.1 |
ENST00000484554.1 |
Ensembl |