Assay Details
| Gene Symbol: RAB34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142624.2 | NP_001136096.2 | 6 - 7 | 554 | 84 | |
| NM_001142625.2 | NP_001136097.2 | 6 - 7 | 557 | 84 | ||
| NM_001144942.1 | NP_001138414.1 | 5 - 6 | 1006 | 84 | ||
| NM_001144943.1 | NP_001138415.1 | 6 - 7 | 554 | 84 | ||
| NM_001256276.1 | NP_001243205.1 | 5 - 6 | 622 | 84 | ||
| NM_001256277.1 | NP_001243206.1 | 6 - 7 | 688 | 84 | ||
| NM_001256278.1 | NP_001243207.1 | 4 - 5 | 383 | 84 | ||
| NM_031934.5 | NP_114140.4 | 5 - 6 | 1006 | 84 | ||
| GenBank mRNA | AF322067.1 | - | 5 - 6 | 382 | 84 | |
| AF327350.1 | - | 6 - 7 | 440 | 84 | ||
| AJ277106.1 | - | 6 - 7 | 593 | 84 | ||
| AK027312.1 | - | 6 - 7 | 587 | 84 | ||
| AK074689.1 | - | 5 - 6 | 984 | 84 | ||
| AK297850.1 | - | 6 - 7 | 462 | 84 | ||
| AK304633.1 | - | 5 - 6 | 537 | 84 | ||
| BC016841.1 | - | 5 - 6 | 864 | 3902651 | 84 | |
| BC066904.1 | - | 6 - 7 | 603 | 5260472 | 84 | |
| BC091510.1 | - | 5 - 6 | 548 | 30512024 | 84 | |
| BG749591.1 | - | 4 - 5 | 309 | 84 | ||
| BT006702.1 | - | 5 - 6 | 382 | 84 | ||
| CB127976.1 | - | 5 - 6 | 521 | 84 | ||
| CX788239.1 | - | 6 - 7 | 447 | 84 | ||
More Information
Set Membership: |
|

RAB34
Genomic Map