| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000331427.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000580223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
ENST00000584711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008874.hg.1 |
NM_178160.2 |
Refseq |