Assay Details
| Gene Symbol: ANKRD20A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092114.1 | - | 2 - 3 | 256 | 77 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000582468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000431648.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000580995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581181.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000581935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000582468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | ENST00000583348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009218.hg.1 | NR_040113.1 | Refseq |

ANKRD20A5P
Genomic Map