Assay Details
| Gene Symbol: DUX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_137167.1 | 3 - 4 | 1439 | 80 | ||
| GenBank mRNA | HQ266765.1 | - | 2 - 3 | 156 | 80 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
negative regulation of cell proliferation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of G0 to G1 transition
transcription regulatory region sequence-specific DNA binding
protein binding
transcription, DNA-templated
apoptotic process
negative regulation of cell proliferation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of G0 to G1 transition
transcription regulatory region sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000633967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012881.hg.1 | ENST00000633967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000616166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000616166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000616219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | ENST00000616219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000564366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000633967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012881.hg.1 | ENST00000633967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000565211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000569241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000570263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000616166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000616166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000616219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | ENST00000616219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001293798.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012880.hg.1 | NM_001306068.2 | Refseq |

DUX4
Genomic Map