Assay Details
Gene Symbol: SEC11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK311213.1 | - | 1 - 2 | 160 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
