Assay Details
Gene Symbol: HIST1H4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC126276.1 | - | 1 | 117 | 8991992 | 150 |
BC126278.1 | - | 1 | 117 | 8991994 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126276.1, BC126278.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
