Assay Details
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163558.2 | NP_001157030.1 | 1 - 2 | 87 | 128 | |
XM_011514753.2 | XP_011513055.1 | 1 - 2 | 131 | 128 | ||
GenBank mRNA | GQ305133.1 | - | 1 - 2 | 87 | 128 | |
M58594.1 | - | 1 - 2 | 87 | 128 | ||
X54393.1 | - | 1 - 2 | 40 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the GQ305133.1, M58594.1, NM_001163558.2, X54393.1, XM_011514753.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
female pregnancy
lactation
cell proliferation
regulation of multicellular organism growth
cellular protein metabolic process
positive regulation of JAK-STAT cascade
JAK-STAT cascade involved in growth hormone signaling pathway
prolactin receptor binding
hormone activity
protein binding
cell surface receptor signaling pathway
female pregnancy
lactation
cell proliferation
regulation of multicellular organism growth
cellular protein metabolic process
positive regulation of JAK-STAT cascade
JAK-STAT cascade involved in growth hormone signaling pathway
prolactin receptor binding
hormone activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000306482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000615510.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | ENST00000617911.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_000948.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011037.hg.1 | NM_001163558.2 | Refseq |
