Assay Details
| Gene Symbol: RPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289111.1 | NP_001276040.1 | 9 | 3035 | 104 | |
| NM_032194.2 | NP_115570.1 | 10 | 3073 | 104 | ||
| XM_017011354.1 | XP_016866843.1 | 7 | 3509 | 104 | ||
| GenBank mRNA | AK056264.1 | - | 1 | 1520 | 104 | |
| AL833613.1 | - | 9 | 3002 | 104 | ||
More Information
Set Membership: |
|

RPF2
Genomic Map