Assay Details
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC041589.1 | - | 2 - 3 | 208 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
