| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
ENST00000409061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006636.hg.1 |
NM_001271700.1 |
Refseq |