| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000464464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
ENST00000467458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009719.hg.1 |
NR_038926.1 |
Refseq |